Learn More
Medchemexpress LLC Dynapyrazole-A | 2226517-75-3 | MFCD31697735 | 98.2% | 486.69 g/mol | C20H12ClIN4O | 5 MG

Supplier: Medchemexpress LLC HY1318735MG
Dynapyrazole-A is a small-molecule inhibitor used in research to inhibit microtubule-stimulated dynamin ATPase activity and probe dynamin-dependent cellular processes. The compound is supplied as a high-purity powder with recommended solvent and storage conditions for stability.
- Specific inhibitor of microtubule-stimulated dynamin ATPase activity.
- High purity, approximately 98.2% by HPLC.
- Recommended solvent: DMSO, typical concentration 25 mg/mL.
- Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
- Available in small research pack sizes suitable for biochemical assays.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.