Learn More
Abcam Dynasore, Dynamin Inhibitor, Solid, >99% Purity, 50 mg
Description
- Molecular weight: 322.3 Da
- Purity: >99%
- Form: Solid
- Function: Cell-permeable dynamin inhibitor that blocks endocytosis
- Mechanism: Non-competitively inhibits dynamin1, dynamin2 and mitochondrial dynamin GTPase activity
- Selectivity: Selective over other small GTPases
- Storage: Store at -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Form | Solid |
| Target Molecule | Dynamin1, Dynamin2, Mitochondrial Dynamin |
| Quantity | 50 mg |
| For Use With (Application) | Endocytosis inhibition, GTPase activity inhibition |
| Storage Requirements | Store at -20°C. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.