Learn More
DYNC1LI1 Mouse anti-Human, Clone: OTI1C5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501630

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Acts as one of several non-catalytic accessory components of the cytoplasmic dynein 1 complex that are thought to be involved in linking dynein to cargos and to adapter proteins that regulate dynein function. Cytoplasmic dynein 1 acts as a motor for the intracellular retrograde motility of vesicles and organelles along microtubules. May play a role in binding dynein to membranous organelles or chromosomes. Probably involved in the microtubule-dependent transport of pericentrin. Is required for progress throuh the spindle assembly checkpoint. The phosphorylated form appears to be involved in the selective removal of MAD1L1 and MAD1L2 but not BUB1B from kinetochores.Specifications
DYNC1LI1 | |
Monoclonal | |
Unconjugated | |
DYNC1LI1 | |
DLC-A, DNCLI1, LIC1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
51143 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1C5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9Y6G9 | |
DYNC1LI1 | |
Full length human recombit protein of human DYNC1LI1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.