Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dynein Monoclonal Antibody (74.1), eBioscience™

Catalog No. p-9090006
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-977-280 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-977-280 Supplier Invitrogen™ Supplier No. 14977280
Only null left
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

The monoclonal antibody 74.1 recognizes all known forms of the 74 kDa intermediate chain subunit of mammalian cytoplasmic dynein. Cytoplasmic dynein is a microtubule-stimulated ATPase responsible for movement of membrane-bound organelles in a directed manner toward the minus end of microtubules, commonly found at the centrosome. Dynein is responsible for the transport of lysosomes, endosomes, phagosomes, peroxisomes, mitochondria, and vesicles from the endoplasmic reticulum destined for the Golgi. During mitosis, dynein is responsible for positioning the spindle apparatus as well as being localized to the kinetochores to facilitate chromosome movement.The 74.1 antibody recognizes human, mouse, rat, bovine, sheep, Xenopus, porcine, and Drosophila dynein intermediate chain. This 74.1 antibody has been tested by immunocytochemistry of methanol-fixed and permeabilized cells and can be used at less than or equal to 1 μg/mL. This 74.1 antibody has been tested by western blot of reduced and non-reduced HeLa cell lysate and can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

The directed movement of membrane-bounded organelles is an important process for various cellular functions including membrane transport, secretion, and axonal transport. Cytoplasmic dynein, a ubiquitous minus end-directed microtubule-based motor protein, is believed to be responsible for the retrograde transport of membranous organelles from the synapse to the cell body in fast axonal transport. The cytoplasmic dynein complex is composed of two ~530 kDa heavy chains, and intermediate chains of ~74 kDa and ~53-59 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dynein
Applications ELISA, Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 74.1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% Sodium Azide; pH 7.2
Gene DYNLL1
Gene Accession No. P61285, P63167, P63168, P63170, Q3MHR3, Q78P75, Q96FJ2, Q9D0M5
Gene Alias 1700064A15Rik; 6720463E02Rik; 8 kDa dynein light chain; 8 kDa dynein light chain b; 8kD LC; 8kDa LC; 8kDLC; C87222; CDLC2; cytoplasmic dynein light polypeptide; cytoplasmic dynein light polypeptide 1; cytoplasmic light-chain dynein; cytosolic DH IC-1; DLC1; Dlc2; DLC8; DLC8b; DNCL1; DNCL1B; Dnclc1; dynein LC8; dynein light chain 1, cytoplasmic; Dynein light chain 2, cytoplasmic; dynein light chain LC8-type 1; Dynein light chain LC8-type 2; dynein light chain-2; dynein, cytoplasmic, light chain 1; dynein, cytoplasmic, light peptide; dynein, cytoplasmic, light polypeptide 1; dynein, light chain, LC8-type 1; dynein, light chain, LC8-type 2; DYNLL1; DYNLL2; HDLC1; LC8; LC8; PIN; LC8a; LOC540369 protein; mPIN; PIN; Protein inhibitor of neuronal nitric oxide synthase; protein inhibitor of nitric oxide synthase; radial spoke 22 homolog; RSPH22; similar to cytoplasmic dynein light chain 1
Gene Symbols DYNLL1, DYNLL2
Host Species Mouse
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100157773, 100512860, 101118902, 101121274, 140734, 140735, 404151, 540369, 56455, 58945, 68097, 8655
Target Species Human, Mouse, Rat, Bovine, Sheep, Pig, Drosophila, Frog
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.