Learn More
Julabo USA Inc DYNEO DD-1200F R1270 EA 230V
SDP
Supplier:  Julabo USA Inc 9021717.N1.33
Encompass_Preferred
Refrigerated/heating circulator, JULABO, DYNEO DD-1200F, 230V/60Hz, R1270 Refrigerant; -50…+200C; 2kW heater. Cooling power: 1.24kW at 20C; 1.09kW at 0C; 620W at -20C; 370W at -30C; 200W at -40C. Adjustable flow pump 8-27L/min, 1.5-10 PSI. USB communication and USB data-logging. External Pt100 probe port. Color TFT screen, programmable, multi-lingual. M16x1 pump fittings with 8 and 12mm barbs. 5-7.5L bath tank. 13 x 18.5 x 27.6 in, 93lbs
Catalog No. NC2751030
Only null left
Add to Cart
Request a Quote
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov