Learn More
Julabo USA Inc Refrigerated/heating circulator, JULABO, DYNEO DD-310F, 115V/60Hz, R449A Refrigerant. -30…+200C. 1kW heater. Cooling 310W at 20C. Pump 8-27LPM 1.5-10 PSI. USB. Pt100 port. Programmable. M16x1 fittings w 8/12mm barbs. 3-4L bath tank
SDP
Supplier:  Julabo USA Inc 9021713.S1.02
Encompass_Preferred
Refrigerated/heating circulator, JULABO, DYNEO DD-310F, 115V/60Hz, R449A Refrigerant; -30…+200C; 1kW heater. Cooling power: 300W at 20C; 270W at 0C; 210W at -10C; 120W at -20C; 20W at -30C. Adjustable flow pump 8-27L/min, 1.5-10 PSI. USB communication and USB data-logging. External Pt100 probe port. Color TFT screen, programmable, multi-lingual. M16x1 pump fittings with 8 and 12mm barbs. 3-4L bath tank. 9.1 x 15.7 x 25.6 in, 60lbs
Catalog No. 50-224-6232
Only null left
Add to Cart
Request a Quote
This item is not returnable. View return policy
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.