Learn More
Bachem Dynorphin A (1-13)
SDP
Supplier:  Bachem 4004062.0001
Encompass_Preferred
Dynorphin A (1-13), H-Tyr-Gly-Gly-Phe-Leu-Arg-Arg-Ile-Arg-Pro-Lys-Leu-Lys-OH, 72957-38-1, C75H126N24O15, Mr 1603.98, 1mg. Dynorphin A (1-13), porcine. Endogenous kappa-opioid receptor agonist. It is suggested that it attenuates galanin-induced impairment of memory processes through the mediation of kappa-opioid receptors.
Catalog No. 50-259-635
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.