Learn More
CPC Scientific H-Tyr-Gly-Gly-Phe-Leu-Arg-Arg-Ile-Arg-Pro-Lys-Leu-Lys-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific DYNP008A
Encompass
ONE-LETTER SEQUENCE: YGGFLRRIRPKLK-NH2MOLECULAR FORMULA: C75H127N25O14MOLECULAR WEIGHT:1602.97STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [79515-34-7], SYNONYMS: Dynorphin A (1-13) amideRESEARCH AREA: Pain
Catalog No. 50-210-9215