Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Dynorphin A, Kappa Agonist, 1 mg

Catalog No. 502707607
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7607 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7607 Supplier Abcam Supplier No. AB1204121MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Endogenous potent κ agonist with analgesic and cardiovascular regulatory effects.

  • Molecular weight: 2,147.51 g/mol
  • Biological activity: Endogenous potent κ agonist
  • Key effects: Analgesic effects in vivo; involved in cardiovascular regulation
  • Composition: 100% Dynorphin A

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Content And Storage Store at -20°C.
Buffer 100% Dynorphin A
Format Vial
Form Lyophilized
Shipping Condition Ambient
Target Molecule Dynorphin A
Formulation 100% Dynorphin A
Quantity 1 mg
For Use With (Application) Analgesia, cardiovascular regulation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.