Learn More
Cayman Chemical DYRK1A InhIbItr comp 11 10mg
SDP
Supplier:  Cayman Chemical 4171510 MG
Encompass_Preferred
A PTH1R agonist selectively induces cAMP production in COS-7 cells expressing human PTH1R (EC50 2.4 UM) over COS-7 cells expressing human PTH2R (EC50 100 UM) induces calcium release from isolated fetal rat long bones at 1 and 3 UM increases lumbar spine and proximal femur BMD lumbar spine bone strength serum levels of osteocalcin and bone formation in the lumbar spine without affecting serum calcium levels in an OVX rat model of osteopenia at 10 mg/kg per day increases serum but not urinary calcium levels in a TPTX rat model of hypothyroidism at 9 mg/kg
Catalog No. 50-294-3417
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.