Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DYRK1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA514467
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA514467 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA514467 Supplier Invitrogen™ Supplier No. PA514467
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DYRK1B Polyclonal Antibody for Western Blot, IHC (P)

DYRK1B is a member of the DYRK family of protein kinases and is proposed to be involved in nuclear functions. Integrins are heterodimers comprised of alpha and beta subunits, that are noncovalently associated transmembrane glycoprotein receptors. Integrins mediate cell-matrix or cell-cell adhesion, and transduced signals that regulate gene expression and cell growth. DYRK1B encodes the integrin beta 4 subunit, a receptor for the laminins. Mutations in DYRK1B are associated with epidermolysis bullosa with pyloric atresia. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for DYRK1B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DYRK1B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene DYRK1B
Gene Accession No. Q9Y463, Q9Z188
Gene Alias AOMS3; dual specificity tyrosine phosphorylation regulated kinase 1B; dual specificity tyrosine-(Y)-phosphorylation regulated kinase 1B; dual specificity tyrosine-phosphorylation-regulated kinase 1B; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 1B; DYRK1B; DYRKB; Minibrain-related kinase; Mirk; Mirk protein kinase
Gene Symbols DYRK1B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 561-589 amino acids from the C-terminal region of human DYRK1B.
Purification Method Ammonium Sulfate Precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13549, 9149
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.