Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DYRK3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111727
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111727 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111727 Supplier Invitrogen™ Supplier No. PA5111727
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GVYDTFMMID ETKCPPCSNV LCNPSEPPPP RRLNMTTEQF TGDHTQHFLD GGEMKVEQLF QEFGNRKSNT IQ Highest antigen sequence indentity to the following orthologs: Mouse - 74%, Rat - 75%.

DYRK3 belongs to the DYRK family of dual-specificity protein kinases that catalyze phosphorylation of serine/threonine and tyrosine residues. The members of this family share structural similarity, but differ in their substrate specificity, suggesting their involvement in different cellular functions. DYRK3 is a negative regulator of EPO-dependent erythropoiesis, may place an upper limit on red cell production during stress erythropoiesis. Inhibits cell death due to cytokine withdrawal in hematopoietic progenitor cells. May act by regulating CREB/CRE signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DYRK3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DYRK3
Gene Accession No. O43781
Gene Alias BC006704; dual specificity tyrosine phosphorylation regulated kinase 3; dual specificity tyrosine-(Y)-phosphorylation regulated kinase 3; dual specificity tyrosine-phosphorylation-regulated kinase 3; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 3; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 5; dyrk3; dyrk3 {ECO:0000312; DYRK5; EMBL:AAH97474.1}; hYAK3-2; KSR; protein kinase Dyrk3; RED; REDK; regulatory erythroid kinase; RP11-343H5.2; RSU2
Gene Symbols DYRK3
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human DYRK3. Recombinant protein control fragment (Product #RP-108031).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8444
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.