Learn More
Aviva Systems Biology Corp DYSF Antibody - middle region (ARP45479_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45479_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DYSF. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DYSF. Alternate names: FER1L1, FLJ00175, FLJ90168, LGMD2B, MMD1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59038
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.