Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dysprosium(III) chloride, ultra dry, 99.98% (REO)

Catalog No. AA3569206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3569206 5 g
AA3569214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3569206 Supplier Thermo Scientific Chemicals Supplier No. 03569206
Add to Cart

CAS: 10025-74-8 | Cl3Dy | 268.85 g/mol

It is used as a precursor for the preparation of other dysprosium salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-74-8
Molecular Formula Cl3Dy
Molecular Weight (g/mol) 268.85
MDL Number MFCD00010984
InChI Key BOXVSFHSLKQLNZ-UHFFFAOYSA-K
IUPAC Name dysprosium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Dy+3]

Specifications

Melting Point 718°C
Physical Form Powder
Quantity 5 g
Merck Index 14,3482
Solubility Information Soluble in water.
Formula Weight 268.86
Percent Purity 99.98%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 3.67 g/mL
Assay (REO)
Chemical Name or Material Dysprosium(III) chloride, Ultra dry
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-039-9
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.