Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DZIP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. p-7206051 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-9001 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-9001 Supplier Proteintech Group Inc Supplier No. 137791AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May participate in spermatogenesis via its interaction with DAZ.

Specifications

Antigen DZIP1
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DZIP1
Gene Accession No. Q86YF9, Q8BMD2
Gene Alias DAZ interacting protein 1, DAZ interacting protein 1/2, DZIP, DZIP1, DZIP2, DZIPt1, KIAA0996, RP11 23E3.3, Zinc finger protein DZIP1
Gene Symbols Dzip1
Host Species Rabbit
Immunogen DZIP1 Fusion Protein Ag4772
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22873, 66573
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.