Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(E)-3-[3-(4-Fluorophenyl)-1-isopropylindol-2-yl]acrolein 98.0+%, TCI America™
SDP

Catalog No. F081925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F081925G 25 g
F08195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F081925G Supplier TCI America Supplier No. F081925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 93957-50-7
Molecular Formula C20H18FNO
Molecular Weight (g/mol) 307.368
MDL Number MFCD03840863
InChI Key DVWHSTKQJBIYCK-VMPITWQZSA-N
Synonym (E)-3-[3-(4-Fluorophenyl)-1-isopropylindol-2-yl]propenal
PubChem CID 9926593
IUPAC Name (E)-3-[3-(4-fluorophenyl)-1-propan-2-ylindol-2-yl]prop-2-enal
SMILES CC(C)N1C2=CC=CC=C2C(=C1C=CC=O)C3=CC=C(C=C3)F

Specifications

Melting Point 131°C
Color Yellow
Quantity 25 g
UN Number 3077
Formula Weight 307.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (E)-3-[3-(4-Fluorophenyl)-1-isopropylindol-2-yl]acrolein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.