Learn More
Medchemexpress LLC HY-15551 10mg , E-4031 CAS:113559-13-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1555110MG
Encompass_Preferred
HY-15551 10mg E-4031 CAS: 113559-13-0 E-4031 is a selective hERG potassium channel blocker for use in class III anti-arrhythmic studies. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0169
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.