Learn More
Selleck Chemical LLC E-4031 dihydrochloride-5mg
SDP
Supplier:  Selleck Chemical LLC S66275MG
Encompass_Preferred
E-4031 is an experimental class III antiarrhythmic drug that blocks potassium channels of the hERG-type.
Catalog No. 50-238-9893