Learn More
Abcam E-4031 Dihydrochloride, HERG K+ Channel Blocker, 99% Purity, Solid, 5 mg
Description
- Molecular Weight: 474.4 Da
- Purity: >99%
- Physical State: Solid
- Activity: Type III antiarrhythmic drug that blocks ion channels encoded by the ether-a-go-go related gene (ERG1 or KCNH1). Blocks channels in the open configuration with little effect on the channels in the closed configuration. The IC50 for block of human ERG channels in expression systems is approx. 350 nM.
- Storage: Store at +4°C.
- Constituents: 100% N-(4-{1-[2-(6-methylpyridin-2-yl)ethyl]piperidine-4-carbonyl}phenyl)methanesulfonamide dihydrochloride
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | HERG K+ Channel |
| Substrate Properties | Chemical Substrate |
| Shipping Condition | Ambient |
| Format | Vial |
| Product Type | Antioxidant |
| Form | Solid |
| Quantity | 5 mg |
| For Use With (Application) | Ion channel blocking |
| Storage Requirements | Store at 4°C. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.