Learn More
Abcam E-4031 Dihydrochloride, HERG K+ Channel Blocker, Solid, 10 mg
Description
- Molecular Weight: 474.4 Da
- Purity: >99%
- Form: Solid
- Description: Type III antiarrhythmic drug that blocks ion channels encoded by the ether-a-go-go related gene (ERG1 or KCNH1).
- Mechanism of Action: Blocks channels in the open configuration with little effect on the channels in the closed configuration.
- IC50: Approx. 350 nM for block of human ERG channels in expression systems.
- Constituents: 100% N-(4-{1-[2-(6-methylpyridin-2-yl)ethyl]piperidine-4-carbonyl}phenyl)methanesulfonamide dihydrochloride
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | HERG K+ Channel |
| Form | Solid |
| Shipping Condition | Ambient |
| Product Type | Antioxidant |
| Content And Storage | Store at 4°C. |
| Formulation | 100% N-(4-{1-[2-(6-methylpyridin-2-yl)ethyl]piperidine-4-carbonyl}phenyl)methanesulfonamide dihydrochloride |
| Quantity | 10 mg |
| For Use With (Application) | HERG K+ channel blockade |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.