Description
- Ideal for smaller ear canals
- Poly bag keeps each pair of ear plugs clean before use
- Test compatible with 3M™ E-A-Rfit™ Validation System
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
Specifications
| ANSI S3.1974 | Yes |
| CSA Z94.2 | Class AL |
| Size | Small |
| For Use With (Application) | Assembly, Blasting, Cleaning, Demolition, Electrical, Facility Maintenance, Grinding, Machine Operations, Painting, Sanding, Welding |
| High Visibility | Yes |
| Model | 312-1208 |
| Packaging Type | Poly bag |
| Pairs per Package | 2000 Pairs/Cs |
| Plug Material | Foam |
| Plug Color | Yellow |
| Show More |