Description
- Soft foam stem for secure grip
- No roll down helps keep ear tip clean
- Ideal for workers with gloved or dirty hands or for situations when only one hand is free
- Compatible with the 3M™ E-A-Rfit™ Dual-ear Validation System
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
Specifications
| ANSI S3.1974 | Yes |
| CSA Z94.2 | Class AL |
| Size | Regular |
| For Use With (Application) | Assembly, Blasting, Cleaning, Demolition, Facility Maintenance, Grinding, Machine Operations, Painting, Paint Preparation, Sanding, Welding |
| High Visibility | Yes |
| Model | 328-1000 |
| Packaging Type | Poly bag |
| Pairs per Package | 100 Pairs/Pk. |
| Plug Material | Elastomeric polymer |
| Plug Color | Yellow |
| Show More |