Description
- No roll-down, easy-insertion design keeps tip clean
- Push-to-fit stem flexes sideways if impacted during wear
- Officially licensed hearing protection of NASCAR combines style with function
- Cylindrical shape fits most ears comfortably
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
Specifications
| ANSI S3.1974 | Yes |
| CSA Z94.2 | Class AL |
| Size | Universal |
| For Use With (Application) | Assembly, Blasting, Cleaning, Demolition, Electrical, Facility Maintenance, Grinding, Machine Operations, Painting, Sanding, Welding |
| High Visibility | No |
| Model | P1400 |
| Packaging Type | Poly bag |
| Pairs per Package | 400 Pairs/Cs |
| Plug Material | Foam, thermoplastic |
| Plug Color | Gray |
| Show More |