Learn More
3M™ E-A-Rfit™ Probed Test Ear Plugs
Catalog No. 19122219 Shop All 3M Products
Change view
Click to view available options
Model:
393-2000-50
393-2001-50
393-2002-50
393-2003-50
393-2004-50
393-2005-50
393-2006-50
393-2007-50
393-2008-50
393-2009-50
393-2010-50
393-2011-50
393-2012-50
393-2013-50
393-2026-50
Size:
Large
Regular
Small
Universal
Product Type:
Classic™ Plus
Classic™ Regular
Classic™ Small
E-Z-Fit™
Express™
FX™
Flexible Fit
Model 1100
No-Touch™
Push-Ins™
Skull Screws™
TaperFit™
Tri-Flange™
UltraFit™
Yellow Neons™
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Size Product Type
19-122-219 393-2008-50 Universal Express™
19-122-218 393-2007-50 Small Classic™ Small
19-122-214 393-2003-50 Regular Classic™ Regular
19-122-220 393-2009-50 Large Classic™ Plus
19-122-216 393-2005-50 Small E-Z-Fit™
19-122-215 393-2004-50 Universal FX™
19-122-221 393-2010-50 Universal Model 1100
19-122-751 393-2013-50 Universal No-Touch™
19-122-213 393-2002-50 Universal Push-Ins™
19-122-223 393-2012-50 Universal Skull Screws™
19-122-222 393-2011-50 Universal Tri-Flange™
19-122-212 393-2001-50 Universal UltraFit™
19-122-211 393-2000-50 Regular Yellow Neons™
19-072-225 393-2026-50 Universal Flexible Fit
19-122-217 393-2006-50 Regular TaperFit™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Catalog No. 19-122-219 Supplier 3M™ Supplier No. 7000052879
Add to Cart
Add to Cart
This item is not returnable. View return policy
Test ear plugs are modified with a probe tube for use during ear plug fit testing with the 3M™ E-A-Rfit™ Validation System
  • Science-based, quantitative testing gives accurate reports of hearing protection
  • Fast, clear, accurate results are obtained in just seconds
  • Test ear plugs are modified with a probe tube for use during ear plug fit testing with the 3M™ E-A-Rfit™ Validation System
  • 50 pair per case
  • 3M PSD products are for occupational use only
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
TRUSTED_SUSTAINABILITY
Specifications
Size Universal
For Use With (Application) Health & Safety Fit Testing
High Visibility No
Model 393-2008-50
Packaging Type Poly bag
Pairs per Package 50 Pairs/Cs
Plug Material Polyurethane
Plug Color Multi-color
Corded Uncorded
Plug Shape Pod
Product Type Express™
Metal Detectable No