Learn More
Medchemexpress LLC (E)-Tyrphostin AG 556 | 133550-41-1 | >98.0% | 336.39 g·mol⁻¹ | C20H20N2O3 | 100 MG
Supplier: Medchemexpress LLC HY101041100MG
(E)-AG 556 is a selective small-molecule inhibitor of the epidermal growth factor receptor (EGFR) tyrosine kinase used in cell signaling and pharmacology research. It has been shown to inhibit EGFR-dependent signaling and to block lipopolysaccharide (LPS)-induced tumor necrosis factor-alpha (TNF-α) production in cellular assays. Supplied as a purified solid, it is typically prepared as DMSO stock solutions for in vitro experiments.
- Selective inhibitor of EGFR tyrosine kinase.
- Inhibits LPS-induced TNF-α production in cell assays.
- High purity suitable for in vitro research applications.
- Soluble in DMSO for concentrated stock preparation.
- Provided as a solid powder for laboratory use.