Learn More
Medchemexpress LLC (E)-AG 556 (E)-Tyrphostin AG 556 | 133550-41-1 | 99.9% | 336.39 g/mol | C20H20N2O3 | 50 MG
Supplier: Medchemexpress LLC HY10104150MG
(E)-AG 556 is a small-molecule research compound that selectively inhibits the epidermal growth factor receptor (EGFR) and reduces LPS-induced tumor necrosis factor-alpha (TNF-α) production. It is supplied as a high-purity solid intended for biochemical and cellular studies of EGFR signaling and inflammation pathways.
- Highly selective EGFR inhibitor.
- Reduces LPS-induced TNF-α production in cellular assays.
- High purity suitable for research applications.
- Characterized chemical with CAS 133550-41-1 for unambiguous identification.
- Supplied in a small-pack solid form for laboratory use.