Learn More
Medchemexpress LLC (E)-Tyrphostin AG 556 | 133550-41-1 | 99.9% | 336.39 g/mol | C20H20N2O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1010415MG
Encompass_Preferred
(E)-AG 556 is a small-molecule epidermal growth factor receptor (EGFR) inhibitor that also blocks LPS-induced TNF-α production. Supplied as a light yellow to yellow solid with high purity, it is used in biochemical and cell-based assays to investigate EGFR signaling and inflammation-related pathways.
  • Selective EGFR inhibitor for signaling studies.
  • Blocks LPS-induced TNF-α production, supporting inflammation research.
  • High purity suitable for in vitro assays.
  • Available as solid and DMSO solution for flexible use.
  • Light yellow to yellow solid for easy visual identification.
Catalog No. 50-002-11176
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.