Learn More
BIOSEARCH TECHNOLOGIES INC (Lucigen) E. cloni 10GF' ELITE Electrocompetent Cells, 24 Rxns (DUOs)Clone, construct libraries, transform Gibson Assembly reactions all with high-yield, T1 phage-resistant cells in high efficiencies
Supplier:  BIOSEARCH TECHNOLOGIES INC 600612
Encompass_Preferred
Direct replacements for standard cloning strainsOptimized genetics for high yields: phage T1 resistant, endonuclease and recombination minus, blue/white screening-capable.Available in a range of high transformation efficienciesConvenient packaging options.Competent Cells include Control DNA and Recovery Medium, and are packaged as SOLOs (1 transformation per tube), DUOs (2 transformations per tube), Subcloning Grade (12 transformations per tube), or microplates as indicated. Recovery Medium is also available separately. The specified transformation efficiencies are with pUC DNA, unless indicated otherwise.
Catalog No. NC2160359
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.