Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ E. coli anti-pan-ADP-ribose binding reagent, Polyclonal,

Catalog No. MABE1016MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE1016MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE1016MI Supplier MilliporeSigma™ Supplier No. MABE1016
Add to Cart
Add to Cart

E. coli Polyclonal Antibody

Specifically detects pan-ADP-ribose binding reagent in Human, Mouse samples, and it is validated for Dot Blot

Anti-pan-ADP-ribose binding reagent, is a His-tagged recombinant protein fused to rabbit Fc tag, expressed in and purified from Rosetta™(DE3)pLysS strain of E. coli. Anti-pan-ADP-ribose binding reagent is useful for the affinity detection of both mono- and poly-ADP-ribosylated proteins on membranes in a manner similar to antibody-based Western and dot blot analysis, The rabbit Fc tag allows visualization of the binding with conjugated anti-rabbit secondary antibodies. The Fc tag also allows Anti-pan-ADP-ribose binding reagent to be captured on Protein A resins for affinity pull-down applications.
TRUSTED_SUSTAINABILITY

Specifications

Antigen pan-ADP-ribose binding reagent
Applications Dot Blot
Classification Polyclonal
Formulation Purified from E. coli by Ni-NTA agarose. Supplied in buffer containing 10mM Tris pH 7.5, 0.2M NaCl, 10% Glycerol, 10mM Imidazole, 1mM PMSF, 1mM β-Mercaptoethanol, 10% glycerol without preservatives.
Host Species E. coli
Purification Method Ni-NTA agarose
Quantity 200 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Stable for one year at -80°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -80°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.