Learn More
Sino Biological E. coli BirA PAb 20ul (Sino Biological)
Supplier:  Sino Biological 11582T1620
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant E. coli BirA / Bifunctional protein BirA (Catalog#11582-E10E; P06709; Met1-Lys321). BirA / Bifunctional protein BirA specific IgG was purified by E. coli BirA / Bifunctional protein BirA affinity chromatography.
Catalog No. 50-237-3906
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.