Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E. coli Exonuclease I, 5000 U, G-Biosciences
SCI_ED

Catalog No. S001141
Change view
Click to view available options
Form:
Liquid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
S001141 Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001141 Supplier G-Biosciences Supplier No. 7862268
Add to Cart
Add to Cart

Exonuclease I (Exo I), is a highly processive single-stranded DNA-specific exonuclease which removes mono- and dinucleotides from the 3' end of single-stranded DNA molecules (3' to 5' exonuclease activity) leaving double-stranded molecules and the 5' end of single stranded molecules intact.

Applications:

  • Removes primer from PCR mixtures
  • PCR mutagenesis
  • Removes single-stranded DNA containing a 3'-hydroxyl terminus from nucleic acid mixtures

Specifications

Packaging Vial
Enzyme Exonuclease
Sensitive to Heat Inactivation Yes
Quantity 250 mL
Product Type Exonuclease I
Research Category Traditional Cloning, DNA Repair
Product Line Thermo Scientific
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.