Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E. coli Gyrase, 100 U, G-Biosciences
SCI_ED

Catalog No. S001161
Change view
Click to view available options
Form:
Liquid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
S001161 Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001161 Supplier G-Biosciences Supplier No. 7862273
Add to Cart
Add to Cart

Gyrase is a type IA DNA topoisomerase and it is involved in bacterial DNA supercoiling.

  • Gyrase is the only type II isomerase that can introduce negative supercoils into DNA.
  • It is a tetramer composed of two A and two B subunits that form a 370 kDa holoenzyme.

Specifications

Packaging Vial
Enzyme Gyrase
Sensitive to Heat Inactivation Yes
Product Type Gyrase
Research Category Drug Development, DNA Repair
Product Line Thermo Scientific
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.