Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ E. coli J5 LPS Monoclonal Antibody (2D7/1)
GREENER_CHOICE

Catalog No. MA183152 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA183152 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA183152 Supplier Invitrogen™ Supplier No. MA183152
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody cross-reacts with K. pneumoniae and S. sonnei and S. typhimurium. Mouse anti Escherichia coli J5 LPS antibody, clone 2D7/1 recognizes E. coli J5 LPS. The J5 mutant of E. coli lacks the enzyme uridine diphosphate glucose 4-epimerase and therefore produces an incomplete LPS, deficient in galactose and all the sugars distal to the central polymers.

Escherichia coli (E. coli) are Gram negative enteric bacteria that live in the human gut. Its presence in nature is an indication of human feacal pollution. E. coli also causes human disease such as urinary tract infections and neonatal meningitis. Infection with particular enterotoxigenic strains can lead to life threatening intestinal diseases. E. coli are distinguished immunologically by serotyping based on three types of antigen: the somatic or O antigen which corresponds to terminal sugars on the cell surface lipopolysaccharide, the capsular or K antigens and the flagellar or H antigens.
TRUSTED_SUSTAINABILITY

Specifications

Antigen E. coli J5 LPS
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2D7/1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with <0.1% sodium azide
Gene Alias Escherichia coli
Host Species Mouse
Immunogen E. coli J5 cells.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.