Learn More
E. coli Poly(A) Polymerase, 500 U, G-Biosciences
SCI_ED
Catalog No. S001159
Change view
Click to view available options
Promoter:
T7
Catalog No. Promoter
S001159 T7
1 options
Catalog No. S001159 Supplier G-Biosciences Supplier No. 7862275
Add to Cart
Add to Cart
Enzyme that catalyzes the addition of adenosine monophosphate (AMP) residues from adenosine triphosphate (ATP) to the 3' end of RNA molecules, creating a polyadenylated tail.
  • It is useful to generate a Poly A tail.
Specifications
Polymerase Poly(A) RNA Polymerase
Product Line Ambion
Promoter T7