Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E. coli Pyrophosphatase (PPase), 50 U, G-Biosciences
SCI_ED

Catalog No. S001118
Change view
Click to view available options
Form:
Liquid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
S001118 Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001118 Supplier G-Biosciences Supplier No. 7862266
Add to Cart
Add to Cart

E. coli Inorganic Pyrophosphatase (PPase) speeds up the breakdown of inorganic pyrophosphate (PPi) into orthophosphate.

  • This enzyme requires a divalent metal cation, preferably Magnesium.
  • Widely applied in in vitro transcription (IVT) or RNA synthesis reactions, PPase minimizes the inhibitory impact of PPi, ensuring smooth and efficient processes.

Specifications

Packaging Vial
Enzyme Pyrophosphatase
Sensitive to Heat Inactivation Yes
Product Type E. coli Inorganic Pyrophosphatase
Research Category RNA Research, Drug Development, Genome Editing
Product Line Thermo Scientific
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.