Learn More
New England Biolabs, Inc. E.coli Ribosome – 1 mg
SDP
Supplier:  New England Biolabs, Inc. P0763S
Encompass
The E. coli 70S ribosome consists of the large 50S and the small 30S subunits.
Catalog No. 50-443-9
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.