Learn More
Apexbio Technology LLC (E,E)-Bisdemethoxycurcumin 33171-05-0 10mg
Supplier: Apexbio Technology LLC A465210
(E E)-Bisdemethoxycurcumin (CAS 33171-05-0) is a naturally derived curcuminoid and a structural analog within the demethoxycurcumin class Research demonstrates that this compound modulates multiple inflammatory signaling pathways and alters the expression of various inflammatory mediators It has been shown to induce cell cycle arrest promote apoptosis in cancer cells and inhibit proliferation-related pathways (E E)-Bisdemethoxycurcumin is widely utilized in studies investigating the molecular mechanisms of inflammation tumor cell biology and the pharmacological properties of curcuminoid derivatives
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.