Learn More
LIFE TECH (BID ECHO ONLY) E-Gel™ 96 Agarose Gels, 1%, 8 Gels
Supplier:  LIFE TECH (BID ECHO ONLY) G700801
Encompass
Invitrogen E-Gel 96 high-throughput precast agarose gels with or without SYBR Safe DNA gel stain are designed for fast, convenient, and safe DNA sample electrophoresis. Each E-Gel agarose gel cassette contains all components required for efficient gel separation and analysis—just load your samples and run.
Catalog No. NC3859528