Learn More
Invitrogen™ E-Gel™ Go! Adapter for E-Gel™ Power Snap Electrophoresis Device
Catalog No. A33811
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 adapter
Catalog No. Quantity
A33811 1 adapter
1 options
Catalog No. A33811 Supplier Invitrogen™ Supplier No. A33811
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Add to Cart
Includes
1 Invitrogen E-Gel™ Go! Adapter
Adapter enables use of low throughput E-Gel Go! agarose gels on E-Gel Power Snap Electrophoresis device.
The Invitrogen E-Gel Go! Adapter is an accessory item for the E-Gel Power Snap Electrophoresis Device. The adapter enables the use of low-throughput E-Gel Go! agarose gels designed for fast separation of up to four DNA samples.
E-Gel Go! precast agarose gel cassettes are the perfect option for performing agarose gel electrophoresis when you have only a few samples to analyze. E-Gel Go! precast agarose gel cassettes have four wells and are premixed with a blue light-excitable nucleic acid stain. E-Gel Go! precast agarose gel cassettes can be stored at room temperature and are ready to use whenever you need them without additional buffers or reagents.
  • Enable use of low throughput E-Gel Go! agarose gels on E-Gel Power Snap Electrophoresis System
  • Save more by using E-Gel Go! agarose gels designed for 4 sample runs
Warranty and Services
1 year warranty
TRUSTED_SUSTAINABILITY
Specifications
Product Line E-Gel
Type Adapter
No. of Wells Adapter
Quantity 1 adapter
Shipping Condition Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.