Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ E-Gel™ Power Snap Electrophoresis Device Starter Kit, DC Safe 2%

Catalog No. G8172ST
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G8172ST 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G8172ST Supplier Invitrogen™ Supplier No. G8172ST
Save 22% on Invitrogen Gels, Reagents, and More
Use promo code GELS22
View all promotions
Add to Cart
Add to Cart

The Invitrogen™ E-Gel™ Power Snap Electrophoresis Device Starter Kit, DC Safe 2%, provides everything you need to quickly start using precast E-Gel™ Double Comb Agarose Gels with SYBR Safe DNA Gel Stain on the device.

The kit includes:

  • Ten E-Gel™ Double Comb Agarose Gels with SYBR Safe DNA Gel Stain, 2% (Cat. No. A42348)
  • E-Gel™ Power Snap Electrophoresis Device (Cat. No. G8100)
  • E-Gel™ Low Range Quantitative DNA Ladder with E-Gel™ Sample Loading Buffer (1X) (Cat. No. 12373031)
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Ten E-Gel Double Comb Agarose Gels with SYBR Safe DNA Gel Stain, 2% (Cat. No. A42348)
• E-Gel Power Snap Electrophoresis Device (Cat. No. G8100)
• E-Gel Low Range Quantitative DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 12373031)

Store at room temperature.
Type Electrophoresis System Starter Kit
Quantity 1 kit
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.