Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ E-Gel™ Power Snap Electrophoresis Device Starter Kit, SYBR Safe 1%

Catalog No. G8121ST
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G8121ST 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. G8121ST Supplier Invitrogen™ Supplier No. G8121ST
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy

Provides everything needed to quickly start using precast 1% E-Gel agarose gels with SYBR Safe on the device.

The Invitrogen E-Gel Power Snap Electrophoresis Device Starter Kit, SYBR Safe 1%, provides everything you need to quickly start using precast 1% E-Gel agarose gels with SYBR Safe on the device.

The kit includes:

  • 10 E-Gel agarose gels with SYBR Safe, 1% (Cat. No. A42100)
  • E-Gel Power Snap Electrophoresis Device (Cat. No. G8100)
  • E-Gel 1 Kb Plus DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488090)
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 1 electrophoresis device
• 10 E-Gel agarose gels with SYBR Safe, 1%
• 1 vial E-Gel 1 kb Plus DNA Ladder
• 1 power adapter
• 1 pair of viewing glasses

Store at room temperature.

Product Line E-Gel
Type Electrophoresis System Starter Kit
Quantity 1 Kit
Shipping Condition Room Temperature
Max. Voltage 240 VAC

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.