Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ E-Gel™ Power Snap Electrophoresis Device Starter Kit, SizeSelect II 2%

Catalog No. G8162ST
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G8162ST 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G8162ST Supplier Invitrogen™ Supplier No. G8162ST
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Add to Cart

Includes

E-Gel Power Snap Electrophoresis device,orange viewing glasses,E-Gel Go! agarose gel adaptor,region specific cable with power adaptor.

Designed for fast and convenient E-Gel™ agarose gel electrophoresis, from DNA sample loading to gel image. Includes E-Gel™ SizeSelect™ II agarose gel, 2%.

The Invitrogen E-Gel Power Snap Electrophoresis Device Starter Kit with 2% E-Gel SizeSelect II agarose gels provides everything you need for faster and more effective DNA fragment library size selection in a next-generation sequencing (NGS) workflow.

The kit includes:

  • 10 E-Gel SizeSelect II agarose gels, 2% (Cat. No. G661012)
  • E-Gel Power Snap Electrophoresis Device (Cat. No. G8100)
  • E-Gel Sizing DNA Ladder (Cat. No. 10488100)

Warranty and Services

1 year warranty

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 1 electrophoresis device
• 10 E-Gel SizeSelect II gels
• 1 vial E-Gel Sizing DNA Ladder
• 1 power adapter
• 1 pair of viewing glasses

Store at room temperature.
Type Electrophoresis System Starter Kit
Quantity 1 Kit
Shipping Condition Room Temperature
Max. Voltage 48 VDC
Voltage 240 VAC
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.