Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ E-Gel™ Power Snap Plus Electrophoresis Systems with 1-year Standard Warranty

Catalog No. G9311
Encompass_Preferred
Change view
Click to view available options
Type:
E-Gel Camera
Horinzontal Gel Electrophoresis Unit
Horizontal Gel Electrophoresis System
Includes:
Camera only
Camera, Gel, WiFi Dongle
Camera, WiFi Dongle
Electrophoresis device only
Gel Type:
E-Gel 48 1% with SYBR Safe
E-Gel 48 2% with SYBR Safe
E-Gel 96 1% with SYBR Safe
E-Gel 96 2% with SYBR Safe
Not Included
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Includes Gel Type
G9311 Horizontal Gel Electrophoresis System Camera, WiFi Dongle Not Included
G9200 E-Gel Camera Camera only Not Included
G9110 Horinzontal Gel Electrophoresis Unit Electrophoresis device only Not Included
G9331 Horizontal Gel Electrophoresis System Camera, Gel, WiFi Dongle E-Gel 48 1% with SYBR Safe
G9332 Horizontal Gel Electrophoresis System Camera, Gel, WiFi Dongle E-Gel 48 2% with SYBR Safe
G9381 Horizontal Gel Electrophoresis System Camera, Gel, WiFi Dongle E-Gel 96 1% with SYBR Safe
G9382 Horizontal Gel Electrophoresis System Camera, Gel, WiFi Dongle E-Gel 96 2% with SYBR Safe
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. G9311 Supplier Invitrogen™ Supplier No. G9311
  • $6,740.00
    $6,335.65 / Each of 1

    Save $404.35

    6% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

E-Gel Power Snap Plus is an integrated high-throughput E-Gel™ Agarose Electrophoresis System that offers fast, convenient, and streamlined nucleic acid electrophoresis analysis solution.

The all-in-one Invitrogen E-Gel Power Snap Plus Electrophoresis System is designed for fast and convenient high-throughput E-Gel agarose gel electrophoresis, from DNA sample loading to gel imaging. With dry precast E-Gel agarose gel technology, you can run DNA samples in as little as 10 minutes and observe sample separation in real time. It is the only benchtop system that seamlessly integrates high-throughput DNA sample separation and gel imaging into one workflow.

Features of the E-Gel Power Snap Plus Electrophoresis system include:

  • Streamline electrophoresis and image capture with an all-in-one system
  • Compatible with 11- to 96-well E-Gel agarose gels
  • View and capture DNA samples in real-time
  • Access your images anywhere using the built-in Thermo Fisher Connect and network compatibility

Small, fast, and powerful

The all-in-one E-Gel Power Snap Plus Electrophoresis System provides everything you need for DNA gel electrophoresis and gel visualization in a small lightweight benchtop system. The system includes an electrophoresis power supply, blue-light transilluminator, amber filter, and adapter for 11- to 22-well E-Gel agarose gels. This enables gel separation and real-time sample tracking on E-Gel agarose gels pre-stained with SYBR Safe or SYBR Gold II DNA stains. The device arrives with pre-programmed protocols for each type of compatible E-Gel agarose gel.

Instant capture and seamless integration

Capture high-resolution E-Gel agarose gel images with the included easy-to-use E-Gel Power Snap Plus Camera. The camera is directly docked to the E-Gel Power Snap Electrophoresis Device, requiring no external power supply or connection to a desktop computer. The E-Gel Power Snap Plus Camera is also compatible with the included WiFi dongle and has an ethernet port, so you can export your experimental data to the cloud or internal servers for maximum convenience.

Unsurpassed convenience of dry precast E-Gel agarose gel

Run DNA samples in as little as 10 minutes with dry, precast, pre-stained E-Gel agarose gels. The E-Gel dry precast agarose gel technology provides speed and convenience, eliminating any need for gel preparation or staining. E-Gel agarose gels are available in a variety of gel percentage and a choice of two DNA stains. The E-Gel Power Snap Plus Electrophoresis Device is compatible with high-throughput E-Gel 48 and E-Gel 96 gels with SYBR Safe*. A provided adapter extends compatibility to low-throughput E-Gel precast agarose gels, including E-Gel EX, E-Gel SYBR Safe, E-Gel CloneWell II, and E-Gel SizeSelect II gels*.

Safer workflows

The blue light emitted by the E-Gel Power Snap Plus Electrophoresis Device transilluminator ensures maximum possible safety to the user and the samples. A combination of blue light and amber filter also offers real-time viewing of the migration of your samples. The E-Gel Power Snap Plus system is a perfect choice for routine PCR fragment analysis, genotyping, and other applications requiring high-throughput. The compatibility with E-Gel CloneWell II gels also makes it a great choice for a safer cloning workflow with increased efficiency.

Made for convenience

The integrated E-Gel Power Snap Plus Camera (available separately as G9201) touch-screen and on-board software provide an intuitive interface for quick gel image capture and editing. The device also offers built in E-Gel 96 deconvolution into several more readable formats. You can store images in the internal memory of the Power Snap Plus system, on our Thermo Fisher Connect cloud-based platform, or on local network drive to further optimize your workflow.

*The E-Gel Power Snap Plus Electrophoresis System also supports ethidium bromide-stained E-Gel high- and low- throughput agarose gels. The ethidium bromide-stained gels are not suitable for imaging with the E-Gel Power Snap Camera or real-time view.

TRUSTED_SUSTAINABILITY

Specifications

Description E-Gel™ Power Snap Plus Electrophoresis System
Detection Method Fluorescence, blue light
For Use With (Application) Nucleic Acid Analysis
Gel Compatibility E-Gel agarose gels with SYBR dyes
Running Time 8 to 40 min.
Type Horizontal Gel Electrophoresis System
Gel Percentage Not Included
Gel Type Not Included
Includes Camera, WiFi Dongle
Quantity 1 system
Warranty 1-year standard warranty included
Show More Show Less
In the E-Gel Power Snap Plus Electrophoresis System, the matt surface of the transilluminator does not look even. Is this a defect?

No, this is expected from the special treatment of the glass. It will have no effect on the final image.

Is the E-Gel Power Snap Plus Electrophoresis System compatible with E-Gel Go! gels?

The E-Gel Power Snap Plus Electrophoresis System is not compatible with E-Gel Go! Gels.

With the E-Gel Power Snap Plus Electrophoresis System, can the E-gel well adapter be easily taken out and put back in?

Yes. There is plenty of space for fingers to grasp the E-gel well adapter in the instrument for best operation.

Do you sell the WiFi dongle for the E-Gel Power Snap Plus Electrophoresis System as a stand-alone item?

Yes, the WiFi dongle for the E-Gel Power Snap Plus Electrophoresis System is offered separately as Cat. No. A26774.

I'm using the E-Gel Power Snap Plus Electrophoresis System and seem to have forgotten my password? What do I do?

A person holding the admin account on the device can help reset the password. If the admin account is affected and is not linked to the Thermo Connect Platform, this account is lost.

Do I have to create a user profile when initially using the E-Gel Power Snap Plus Electrophoresis Device?

No, you have to create a user profile only if you wish to have a separate profile on the device or link it with the Thermo Fisher Connect platform.

Do you provide the adapter for 11-22 well E-Gels with the E-Gel Power Snap Plus Electrophoresis System?

Yes, the adapter is in a drawer in the electrophoresis device. It extends the compatibility to 11-22-well E-Gel gels as well as CloneWell II, SizeSelect II and E-Gel NGS gels.

How many images can I store on the E-Gel Power Snap Plus Electrophoresis System?

This would depend on the selected image quality but we expect that the E-Gel Power Snap Plus Electrophoresis System will store ~2 months of image data in the internal 64 GB memory.

Do you offer IQ/OQ/PQ service for the E-Gel Power Snap Plus Electrophoresis System?

No. The device runs internal diagnostics protocols every time it is turned on. These can also be initiated by the user.

Can I connect the E-Gel Power Snap Plus Electrophoresis Device to WiFi without the dongle?

No. The WiFi dongle is needed for connection of the E-Gel Power Snap Plus Device to the internet. The dongle is included with the E-Gel Power Snap Plus Electrophoresis System and with all E-Gel Power Snap Plus Electrophoresis starter kits.

Can I connect the E-Gel Power Snap Plus Camera directly to a computer?

No. The E-Gel Power Snap Plus Camera contains everything it needs for independent operation: a large 8'' screen with touch input and an intuitive user interface.

Can the E-Gel Power Snap Plus Camera be directly powered using the power cord that is included in the E-Gel Power Snap Plus Electrophoresis System?

No, the E-Gel Power Snap Plus Camera is powered by the base module and has to be connected to the base module to function. It cannot be connected directly to the power cord.

Can I use the same E-Gel Power Snap Plus Camera with other E-Gel Power Snap Plus Electrophoresis Device base modules?

Yes, the E-Gel Power Snap Plus Camera is powered by the E-Gel Power Snap Plus Electrophoresis Device base module and can be used on any E-Gel Power Snap Plus Electrophoresis Device base module.

Can I use the power cord provided with the E-Gel Power Snap Plus Electrophoresis System with the E-Gel Power Snap Electrophoresis System?

Yes, both the E-Gel Power Snap Plus Electrophoresis System and the E-Gel Power Snap Electrophoresis System use identical power cords.

Can I use ethidium bromide E-Gels with the E-Gel Power Snap Plus Electrophoresis System?

Yes. The E-Gel Power Snap Plus Electrophoresis System is compatible with E-Gels containing ethidium bromide. However, as the ethidium bromide dye is not compatible with the included blue light transilluminator, the experience will not be optimal.

Can I image my E-PAGE gels using the E-Gel Power Snap Plus Camera?

No. The E-Gel Power Snap Plus instrument does not have the required illumination type for E-PAGE gel documentation.

Is the E-Gel Power Snap Plus Electrophoresis System compatible with E-PAGE gels?

Yes, the E-Gel Power Snap Plus Electrophoresis System is compatible with E-PAGE gel protocols. The instrument has them located under the gel type 48/96.

I'm using the E-Gel Power Snap Plus Electrophoresis System, and the USB drive is not connecting to the instrument. What should I do?

Make sure that the USB drive you are using is formatted correctly. The instrument recognizes the FAT32 format file system.

With the E-Gel Power Snap Plus Electrophoresis System, the camera failed to startup on the first time of use. What should I do?

Try to re-dock the instrument one more time. This should fix the initial error.


Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.