Learn More
TECHLAB™ E. HISTOLYTICA II™ Diluent
Catalog No. 21279142
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
21-279-142 1 Ea.
1 options
Catalog No. 21-279-142 Supplier TECHLAB™ Supplier No. D2IVD
Add to Cart
Add to Cart
The E. HISTOLYTICA II™ test is an enzyme immunoassay for the rapid detection of the adhesin of E. histolytica in human fecal specimens. It is indicated for use with fecal specimens from patients with diarrhea or dysentery to determine the presence of E. histolytica gastrointestinal infection.
  • The test can be used for fecal specimens submitted for routine clinical testing from adults or children.
  • Conventional microscopy is not a prerequisite for use of the test.
Specifications
Product Type Diluent
Quantity 1 Ea.
Combinability Non-Combinable
Research Area Immunology