Learn More
SiliCycle™ E-PAK™ Cartridges, SiliaMetS™ AMPA
E-PAK™ Cartridges, SiliaMetS™ AMPA is an aminomethylphosphonic acid ligand conveniently packaged in a cartridge. It is particularly efficient to remove Al, Sb, Ni, La, Co, Fe, Mn, and Zn. E-PAK™ lab cartridges are generally used for processing samples of 50 mL or more.
Supplier:  SiliCycle™ LSR85130B513
Catalog No. 01-182-129
Add to cart
Description
Description
Metals removed:
  • Best scavenger: Al, Ce, Dy, Er, Eu, Gd, Ho, La, Lu, Mn, Nd, Ni, Pm, Pr, Sb, Sm, Tb, Tm, Y, and Yb
  • Good scavenger: Co, Cu, Fe, Mg, and Zn
Additional Information:
  • To facilitate the evaluations of small samples, we can provide loose media and lab scale cartridges on a custom basis.
  • Testing with loose media can be done with samples as small as a few milliliters and is normally done in advance of cartridge testing to identify the formula with the highest capacity to remove contaminants with the least loss of product.
  • E-PAK™ lab cartridges are generally used for processing samples of 50 mL or more.
  • Testing has shown a good correlation between the performance of lab, pilot, and commercial size cartridges.
  • Lab cartridges can be operated with a peristaltic pump or small pressure vessel.
Specifications
Specifications
Lab Scale E-PAK Cartridge
White to yellow
3 Pk.
Box
AMPA
Lower residual metal concentration in Al, Sb, Ni, La, Co, Cu, Fe, Mg and Zn
1 x 5 cm
2 to 9
40 to 63 μm
8 g