Learn More
Agilent Technologies™ E-Plate 16 PET
Catalog No. 300600880
Change view
Click to view available options
Quantity:
6 Plates
6 Plates/Pk., 6 Pk./Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-060-0880 6 Plates/Pk., 6 Pk./Cs.
30-060-0890 6 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 30-060-0880 Supplier Agilent Technologies™ Supplier No. 300600880
Add to Cart
Add to Cart
Observe the cell response in real-time with E-Plate 16 PET.
  • Derive cell kinetics
  • Obtain continuous immune cell killing assay time points during the entire experiment
  • RTCA provides a continuous quantitative readout of proliferation rate, cell number, size/shape, and substrate attachment quality
Specifications
Barcode Includes Barcode
Format Standard
Array 2 x 8
Color Clear, Gold
Disposable Single-use
Material Polystyrene
Volume (Metric) Well 270 ±10 μL
No. of Wells 16
Sterility Non-sterile
Surface Treatment Gold-plated
Well Shape Flat Bottom (F-well)
Quantity 6 Plates/Pk., 6 Pk./Cs.