Learn More
Agilent Technologies™ E-Plate Insert 16
Catalog No. 6465382001
Change view
Click to view available options
Quantity:
6 Plates/Pk.
Catalog No. Quantity
64-653-82001 6 Plates/Pk.
1 options
Catalog No. 64-653-82001 Supplier Agilent Technologies™ Supplier No. 6465382001
Add to Cart
Add to Cart
E-Plate Insert 16 with label-free environment to allow detection of responses due to stimulation of endogenous receptors.
  • Kinetic response profiles may be indicative for specific pathways
  • Real time data provides comprehensive information on cell responses over long time periods
  • E-Plate Insert offers an easy to setup co-culture platform with minimal handling
Specifications
Type Slide Insert
For Use With (Equipment) E-Plate 16 PET
Quantity 6 Plates/Pk.