Learn More
Medchemexpress LLC HY-13013 2mg , (E)-SIS3 CAS:521984-48-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY130132MG
Encompass_Preferred
HY-13013 2mg (E)-SIS3 CAS: 521984-48-5 (E)-SIS3 is a potent and selective inhibitor of Smad3 with an IC50 of 3 μM for Smad3 phosphorylation. (E)-SIS3 inhibits the myofibroblast differentiation of fibroblasts by TGF-β1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0695
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.