Learn More
Medchemexpress LLC (E/Z)-FK-506 26,28-allylic ester rearrangement impurity | 131944-48-4 | MFCD28988714 | >90.0% | 804.02 | C44H69NO12 | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW58588625MG
Encompass_Preferred
(E/Z)-FK-506 26,28-allylic ester rearrangement impurity (FK506-A) is an analytical impurity of the macrolide immunosuppressant tacrolimus, provided as a small-scale reference material for research and analytical laboratories. It is intended for impurity identification, method development, and stability or degradation studies.
Catalog No. 50-002-84143