Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ 17beta-Estradiol-D5 Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco E0611ML
Description
A stable-labeled internal standard for 17β-Estradiol testing or isotope dilution methods by GC/MS or LC/MS for endocrinology, clinical chemistry or diagnostic purposes. Estradiol is a sex hormone prescribed under the trade names Vagifem™, Estring™, Evamist, Elestrin™, Estrace™, and Estrogel™ as a treatment for hypoestrogenism.
Specifications
75-05-8, 221093-45-4 | |
MFCD01075456 | |
100 μg/mL in acetonitrile | |
Ampule | |
2°C (35.6°F) - closed cup |
C18H19O2D5 | |
UN 1648 3 / PGII | |
277.41 | |
1 mL |
Safety and Handling
H332-H319
H312
EINECSNumber : 200-835-2
Recommended Storage : -20°C
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation
Spot an opportunity for improvement?Share a Content Correction